Learn More
Medchemexpress LLC 5'-fluoroindirubinoxime | 861214-33-7 | MFCD18086923 | 98.0% | 295.27 g·mol⁻¹ | C16H10FN3O2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10346410MG
Encompass_Preferred
5'-Fluoroindirubinoxime is an indirubin-derived small molecule that acts as a potent FLT3 kinase inhibitor (IC50 = 15 nM) and is supplied for research use in biochemical and cellular assays.
  • Potent FLT3 inhibitor with IC50 of 15 nM.
  • Molecular formula C16H10FN3O2 and molecular weight 295.27 g·mol⁻¹.
  • Available as solid packs (for example, 5 mg and 10 mg) and as a 10 mM solution in DMSO.
  • Reported purity 98.0% (HPLC).
  • Intended for research applications in biochemical and cellular studies of FLT3 signaling.
Catalog No. 50-002-44840