Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroindole-3-carboxaldehyde, 99%, Thermo Scientific™

Catalog No. AC443350010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443350010 1 g
AC443350050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC443350010 Supplier Thermo Scientific Chemicals Supplier No. 443350010

Chemical Identifiers

CAS 2338-71-8
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00022719
InChI Key YUAJKGBLPVLADK-UHFFFAOYSA-N
Synonym 5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
PubChem CID 259089
IUPAC Name 5-fluoro-1H-indole-3-carbaldehyde
SMILES FC1=CC=C2NC=C(C=O)C2=C1

Specifications

Melting Point 170°C to 172°C
Assay Percent Range 98.5 % min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 163.15
Percent Purity 99%
Chemical Name or Material 5-Fluoroindole-3-carboxaldehyde
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.