Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-Fluoroindole | 399-52-0 | 135.14 | 10 G
SDP

Catalog No. 5000405103
Encompass_Preferred

5-Fluoroindole is a laboratory chemical used for the manufacture of substances. It is stable under recommended storage conditions and should be stored in a cool, well-ventilated area, protected from light and nitrogen.

  • Avoid inhalation, contact with eyes and skin.
  • Avoid dust and aerosol formation.
  • Use only in areas with appropriate exhaust ventilation.
  • Keep container tightly sealed.
  • Keep away from direct sunlight and sources of ignition.
  • Recommended storage temperature: 4°C, protect from light, stored under nitrogen.
  • Appearance: Solid.
  • Melting/freezing point: 45-48°C.
  • Boiling point/range: 258°C at 760 mmHg.
  • Flash point: 110°C.

Catalog No. 50-004-05103 Supplier Medchemexpress LLC Supplier No. HY4015610G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluoroindole is a laboratory chemical used for the manufacture of substances. It is stable under recommended storage conditions and should be stored in a cool, well-ventilated area, protected from light and nitrogen.

  • Avoid inhalation, contact with eyes and skin.
  • Avoid dust and aerosol formation.
  • Use only in areas with appropriate exhaust ventilation.
  • Keep container tightly sealed.
  • Keep away from direct sunlight and sources of ignition.
  • Recommended storage temperature: 4°C, protect from light, stored under nitrogen.
  • Appearance: Solid.
  • Melting/freezing point: 45-48°C.
  • Boiling point/range: 258°C at 760 mmHg.
  • Flash point: 110°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.