Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroindole, 99%

Catalog No. p-7022524
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1534603 1 g
AAA1534606 5 g
AAA1534614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1534603 Supplier Thermo Scientific Chemicals Supplier No. A1534603
Only null left
Add to Cart

CAS: 399-52-0 | C8H6FN | 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-52-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00005671
InChI Key ODFFPRGJZRXNHZ-UHFFFAOYSA-N
Synonym 5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
PubChem CID 67861
ChEBI CHEBI:72818
IUPAC Name 5-fluoro-1H-indole
SMILES FC1=CC=C2NC=CC2=C1

Specifications

Melting Point 42°C to 47°C
Flash Point >110°C (230°F)
Quantity 1 g
Beilstein 112350
Formula Weight 135.14
Percent Purity 99%
Chemical Name or Material 5-Fluoroindole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-917-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.