Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroisatin, 98%

Catalog No. AC367700250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC367700250 25 g
AC367700050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC367700250 Supplier Thermo Scientific Chemicals Supplier No. 367700250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 443-69-6 | C8H4FNO2 | 165.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 443-69-6
MDL Number MFCD00022795
InChI Key GKODDAXOSGGARJ-UHFFFAOYSA-N
Synonym 5-fluoroisatin, 5-fluoroindoline-2,3-dione, 5-fluoro isatin, 5-fluoro-2,3-dihydro-1h-indole-2,3-dione, 5-fluoroisatine, 5-fluoro-2,3-indoledione, 5-fluoroindole-2,3-dione, 1h-indole-2,3-dione, 5-fluoro, 5-fluoro-2,3-indolinedione, 5-fluoro-2,3-indolindione
PubChem CID 236566
IUPAC Name 5-fluoro-1H-indole-2,3-dione
SMILES C1=CC2=C(C=C1F)C(=O)C(=O)N2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 224.0°C to 228.0°C
Color Red
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H4FNO2
Quantity 25 g
Molecular Weight (g/mol) 165.12
Formula Weight 165.12
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Fluoroisatin, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.