Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluoroisobenzofuran-1,3-dione | 319-03-9 | MFCD00191363 | 97.0% | 166.11 g/mol | C8H3FO3 | 10 G
SDP

Catalog No. 5000302948
Encompass_Preferred

5-Fluoroisobenzofuran-1,3-dione (4-fluorophthalic anhydride) is an organic reagent used in life-science research and organic synthesis. Supplied as an off-white to gray solid (CAS 319-03-9), it has a molecular weight of 166.11 g/mol and is provided for research use only. Recommended storage and solvent recommendations support stability and handling for laboratory use.

  • Purity 97.0%.
  • Molecular formula C8H3FO3 and molecular weight 166.11 g/mol.
  • Off-white to gray solid appearance.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or veterinary use.
  • Shipping at room temperature in continental US; may vary elsewhere.

Catalog No. 50-003-02948 Supplier Medchemexpress LLC Supplier No. HYW01534910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluoroisobenzofuran-1,3-dione (4-fluorophthalic anhydride) is an organic reagent used in life-science research and organic synthesis. Supplied as an off-white to gray solid (CAS 319-03-9), it has a molecular weight of 166.11 g/mol and is provided for research use only. Recommended storage and solvent recommendations support stability and handling for laboratory use.

  • Purity 97.0%.
  • Molecular formula C8H3FO3 and molecular weight 166.11 g/mol.
  • Off-white to gray solid appearance.
  • Soluble in DMSO at 100 mg/mL; ultrasonic assistance recommended.
  • Storage: powder -20°C (up to 3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Intended for research use only; not for human or veterinary use.
  • Shipping at room temperature in continental US; may vary elsewhere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.