Learn More
Medchemexpress LLC HY-W016819 100mg Medchemexpress, 5-Fluoroorotic acid CAS:703-95-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016819100MG
Encompass_Preferred
Medchemexpress, HY-W016819 100mg 5-Fluoroorotic acid CAS:703-95-7 5-Fluoroorotic is a selective agent in yeast molecular genetics. 5-Fluoroorotic possesses a well-expressed anticandidal effect close to that of 5-fluorocytosine, as well as moderate antidermatophytal effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3403