Learn More
Research Products International Corp 5-FOA [5-Fluoroorotic acid], 25 Grams
SDP
Supplier:  Research Products International Corp F1050125.0
Encompass_Preferred
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.
Catalog No. 50-488-659