Learn More
Research Products International Corp 5-FOA [5-Fluoroorotic acid], 5 Grams
SDP
Catalog No. 50213307
Encompass_Preferred
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.
Catalog No. 50-213-307 Supplier Research Products International Corp Supplier No. F105015.0
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.