Learn More
Research Products International Corp 5-FOA [5-Fluoroorotic acid], 100 Milligrams
SDP
Catalog No. 50213304
Encompass_Preferred
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.
Catalog No. 50-213-304 Supplier Research Products International Corp Supplier No. F105010.1
Reagent for selection of Orotidine-5'-phosphate decarboxylase mutants of Saccharomyces cerevisiae.