Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluoroorotic acid | 703-95-7 | MFCD00042526 | 99.8% | 174.09 g/mol | C5H3FN2O4 | 1 G
SDP

Catalog No. 5000300629
Encompass_Preferred

5-Fluoroorotic acid (5-FOA) is a fluorinated pyrimidine analogue that inhibits thymidylate synthase and is widely used as a selective agent in yeast molecular genetics. It has reported antimalarial activity and is supplied for research applications.

  • CAS number 703-95-7.
  • Molecular formula C5H3FN2O4; molecular weight 174.09 g/mol.
  • High purity, typically ~99.8%.
  • Solid, light brown to brown appearance.
  • Soluble in DMSO (125 mg/mL); compatible with common in vivo vehicles.
  • Stable as powder at -20°C for extended storage; shorter stability in solution.

Catalog No. 50-003-00629 Supplier Medchemexpress LLC Supplier No. HYW0168191G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluoroorotic acid (5-FOA) is a fluorinated pyrimidine analogue that inhibits thymidylate synthase and is widely used as a selective agent in yeast molecular genetics. It has reported antimalarial activity and is supplied for research applications.

  • CAS number 703-95-7.
  • Molecular formula C5H3FN2O4; molecular weight 174.09 g/mol.
  • High purity, typically ~99.8%.
  • Solid, light brown to brown appearance.
  • Soluble in DMSO (125 mg/mL); compatible with common in vivo vehicles.
  • Stable as powder at -20°C for extended storage; shorter stability in solution.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.