Learn More
Medchemexpress LLC 5-fluoroorotic acid (monohydrate) | 220141-70-8 | MFCD00150658 | 98.0% | 192.10 | C5H5FN2O5 | 500 MG
SDP
Catalog No. 5000300630
Encompass_Preferred
5-Fluoroorotic acid monohydrate is the monohydrate form of 5-fluoroorotic acid, a research reagent that inhibits thymidylate synthase. It is commonly used as a selective agent in yeast molecular genetics and has reported antimalarial activity. Supplied as a high-purity solid for laboratory research applications.
  • Inhibits thymidylate synthase and enables selection in yeast genetics.
  • Supplied as a white to light yellow solid with high purity.
  • Provided as the monohydrate form for consistent stoichiometry.
  • Suitable for laboratory research requiring reliable identity and purity.
  • Packaged in lab-scale quantities for experimental use.
Catalog No. 50-003-00630 Supplier Medchemexpress LLC Supplier No. HYW016819B500MG
5-Fluoroorotic acid monohydrate is the monohydrate form of 5-fluoroorotic acid, a research reagent that inhibits thymidylate synthase. It is commonly used as a selective agent in yeast molecular genetics and has reported antimalarial activity. Supplied as a high-purity solid for laboratory research applications.
  • Inhibits thymidylate synthase and enables selection in yeast genetics.
  • Supplied as a white to light yellow solid with high purity.
  • Provided as the monohydrate form for consistent stoichiometry.
  • Suitable for laboratory research requiring reliable identity and purity.
  • Packaged in lab-scale quantities for experimental use.