Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluoroorotic acid monohydrate | 220141-70-8 | MFCD00150658 | 98.0% | 192.10 g/mol | C5H5FN2O5 | 1 G
SDP

Catalog No. 5000301009
Encompass_Preferred

5-Fluoroorotic acid (monohydrate) is a fluorinated pyrimidine carboxylic acid used in molecular biology and biochemical research as an inhibitor of thymidylate synthase and as a selective agent in yeast genetics. Supplied as a white to light yellow solid, it is intended for research use and has reported antimalarial activity.

  • Inhibits thymidylate synthase, useful for biochemical studies.
  • Selectable agent for yeast genetics and ura3-related assays.
  • Monohydrate solid form for reproducible handling.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 192.10 g/mol and formula C5H5FN2O5 for precise calculations.
  • Recommended storage at -20°C under inert atmosphere to maintain stability.

Catalog No. 50-003-01009 Supplier Medchemexpress LLC Supplier No. HYW016819B1G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluoroorotic acid (monohydrate) is a fluorinated pyrimidine carboxylic acid used in molecular biology and biochemical research as an inhibitor of thymidylate synthase and as a selective agent in yeast genetics. Supplied as a white to light yellow solid, it is intended for research use and has reported antimalarial activity.

  • Inhibits thymidylate synthase, useful for biochemical studies.
  • Selectable agent for yeast genetics and ura3-related assays.
  • Monohydrate solid form for reproducible handling.
  • High purity (98.0%) suitable for analytical and biological assays.
  • Molecular weight 192.10 g/mol and formula C5H5FN2O5 for precise calculations.
  • Recommended storage at -20°C under inert atmosphere to maintain stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.