Learn More
Medchemexpress LLC 5-Fluoroorotic acid monohydrate | 220141-70-8 | MFCD00150658 | 98.0% | 192.10 g/mol | C5H5FN2O5 | 5 G

Supplier: Medchemexpress LLC HYW016819B5G
5-Fluoroorotic acid monohydrate is a research-grade pyrimidine derivative that inhibits thymidylate synthase and is commonly used as a selective agent in yeast molecular genetics and biochemical studies. The monohydrate salt is characterized by a defined molecular formula and high purity, making it suitable for laboratory research applications.
- High purity: 98.0%.
- Monohydrate salt form.
- CAS number 220141-70-8.
- Molecular formula C5H5FN2O5; molecular weight 192.10 g/mol.
- Used for genetic selection and enzyme inhibition studies.
- Available in small research pack sizes such as 5 g.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.