Learn More
Cayman Chemical 5-Fluoropentylindole, Purity: ≥95%, CAS: 1859218-30-6, 10 mg
SDP
Catalog No. 502973172
Encompass_Preferred
The base structure for a variety of synthetic CBs; intended for forensic and research applications
Catalog No. 50-297-3172 Supplier Cayman Chemical Supplier No. 1545410 MG
The base structure for a variety of synthetic CBs; intended for forensic and research applications