Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoropyridine-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F08595G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0859-5G 5 g
F08591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0859-5G Supplier TCI America Supplier No. F08595G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 872041-86-6
Molecular Formula C5H5BFNO2
MDL Number MFCD07368243
InChI Key FVEDGBRHTGXPOK-UHFFFAOYSA-N
PubChem CID 44717403
IUPAC Name (5-fluoropyridin-3-yl)boronic acid

Specifications

Melting Point 236°C
Color White-Yellow
Quantity 5 g
Synonym 5-fluoropyridine-3-boronic acid, 5-fluoropyridin-3-yl boronic acid, 5-fluoropyridin-3-yl-3-boronic acid, 5-fluoropyridine-3-boronicacid, 5-fluoro-3-pyridylboronic acid
SMILES OB(O)C1=CC(F)=CN=C1
Molecular Weight (g/mol) 140.91
Formula Weight 140.91
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.