Learn More
Medchemexpress LLC 5-Fluorouracil-15N2 | 68941-95-7 | 99.0% | C4H3F15N2O2 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY90006S21MG
Encompass_Preferred
5-Fluorouracil-15N2 is the 15N-labeled 5-Fluorouracil. 5-Fluorouracil (5-FU) is an analogue of uracil and a potent antitumor agent. It affects pyrimidine synthesis by inhibiting thymidylate synthetase, thus depleting intracellular dTTP pools. 5-Fluorouracil induces apoptosis and can be used as a chemical sensitizer, and it also inhibits HIV.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-33331