Learn More
Sigma Aldrich Fine Chemicals Biosciences 5-Fluorouracil, 51-21-8, MFCD00006018, 5 g
Catalog No. NC0916606
Encompass
Empirical Formula (Hill Notation): C4H3FN2O2, Molecular Weight: 130.08, ≥99% (HPLC), powder, mp: 282-286 °C, Synonym: 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione.
Catalog No. NC0916606 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. F66275G
Empirical Formula (Hill Notation): C4H3FN2O2, Molecular Weight: 130.08, ≥99% (HPLC), powder, mp: 282-286 °C, Synonym: 2,4-Dihydroxy-5-fluoropyrimidine, 5-FU, 5-Fluoro-2,4(1H,3H)-pyrimidinedione.