Learn More
Medchemexpress LLC HY-90006 5g , 5-Fluorouracil 5-FU CAS:51-21-8 Purity:98%
SDP
Catalog No. 501492172
Encompass_Preferred
Medchemexpress, HY-90006 5g 5-Fluorouracil 5-FU CAS:51-21-8 Formula:C4H3FN2O2 Purity:98% 5-Fluorouracil is a potent antitumor agent that affects pyrimidine synthesis by inhibiting thymidylate synthetase thus depleting intracellular dTTP pools. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2172 Supplier Medchemexpress LLC Supplier No. HY900065G
Medchemexpress, HY-90006 5g 5-Fluorouracil 5-FU CAS:51-21-8 Formula:C4H3FN2O2 Purity:98% 5-Fluorouracil is a potent antitumor agent that affects pyrimidine synthesis by inhibiting thymidylate synthetase thus depleting intracellular dTTP pools. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.