Learn More
Enzo Life Sciences 5-FluorouracIL-(5g). CAS: 51-21-8
SDP
Supplier:  Enzo Life Sciences ALX480099G005
Encompass_Preferred
Potent antitumor agent. Inhibits thymidylate synthetase. Induces p53-dependent apoptosis. Alternative name: 5-FU, 2,4-Dihydroxy-5-fluoropyrimidine, 5-Fluoro-2,4(1H,3H)-pyrimidinedione. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (5mg/ml), dimethyl formamide (5mg/ml), methanol (1mg/ml) or hot water (1mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9329