Learn More
Medchemexpress LLC 5-Fluorouracil (Standard) | 51-21-8 | >99.8% | C4H3FN2O2 | 100 MG
SDP
Catalog No. 5000433048
Encompass_Preferred
5-Fluorouracil (Standard) is the analytical standard of 5-Fluorouracil, intended for research and analytical applications. This product, also known as 5-FU, is an analogue of uracil and a potent antitumor agent. It impacts pyrimidine synthesis by inhibiting thymidylate synthetase, leading to a depletion of intracellular dTTP pools. Additionally, 5-Fluorouracil induces apoptosis and acts as a chemical sensitizer, and has also been observed to inhibit HIV.
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments
  • Applicable in HPLC, GC and MS analysis
Catalog No. 50-004-33048 Supplier Medchemexpress LLC Supplier No. HY90006R100MG
5-Fluorouracil (Standard) is the analytical standard of 5-Fluorouracil, intended for research and analytical applications. This product, also known as 5-FU, is an analogue of uracil and a potent antitumor agent. It impacts pyrimidine synthesis by inhibiting thymidylate synthetase, leading to a depletion of intracellular dTTP pools. Additionally, 5-Fluorouracil induces apoptosis and acts as a chemical sensitizer, and has also been observed to inhibit HIV.
  • Reference standard supplied assay
  • Used in qualitative, quantitative and methodological research experiments
  • Applicable in HPLC, GC and MS analysis