Learn More
Medchemexpress LLC 5-fluorouracil (standard) | 51-21-8 | 99.8% | C4H3FN2O2 | 25 MG
SDP
Catalog No. 5000357521
Encompass_Preferred
5-Fluorouracil (Standard) is the analytical standard of 5-Fluorouracil, an analogue of uracil and a potent antitumor agent. It is intended for research and analytical applications. 5-Fluorouracil affects pyrimidine synthesis by inhibiting thymidylate synthetase, leading to depleted dTTP pools. It induces apoptosis, acts as a chemical sensitizer, and inhibits HIV.
  • Analytical standard for 5-Fluorouracil
  • Suitable for research and analytical applications
  • Analogue of uracil and potent antitumor agent
  • Inhibits thymidylate synthetase and depletes dTTP pools
  • Induces apoptosis and acts as a chemical sensitizer
  • Inhibits HIV
  • Used as a reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-003-57521 Supplier Medchemexpress LLC Supplier No. 25MGHY90006R
5-Fluorouracil (Standard) is the analytical standard of 5-Fluorouracil, an analogue of uracil and a potent antitumor agent. It is intended for research and analytical applications. 5-Fluorouracil affects pyrimidine synthesis by inhibiting thymidylate synthetase, leading to depleted dTTP pools. It induces apoptosis, acts as a chemical sensitizer, and inhibits HIV.
  • Analytical standard for 5-Fluorouracil
  • Suitable for research and analytical applications
  • Analogue of uracil and potent antitumor agent
  • Inhibits thymidylate synthetase and depletes dTTP pools
  • Induces apoptosis and acts as a chemical sensitizer
  • Inhibits HIV
  • Used as a reference standard in assays
  • Commonly employed in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)