Learn More
Medchemexpress LLC 5-Fluorouracil (standard) | 51-21-8 | 99.83% | C4H3FN2O2 | 50 MG
SDP
Catalog No. 5000433081
Encompass_Preferred
5-Fluorouracil (standard) is the analytical standard of 5-Fluorouracil, intended for research and analytical applications. It functions as an analogue of uracil and is recognized as a potent antitumor agent. Its mechanism involves affecting pyrimidine synthesis through the inhibition of thymidylate synthetase, leading to a depletion of intracellular dTTP pools.
  • Induces apoptosis and can serve as a chemical sensitizer.
  • Inhibits HIV.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Applicable in techniques such as HPLC, GC, and MS.
Catalog No. 50-004-33081 Supplier Medchemexpress LLC Supplier No. HY90006R50MG
Add to Cart
Add to Cart
5-Fluorouracil (standard) is the analytical standard of 5-Fluorouracil, intended for research and analytical applications. It functions as an analogue of uracil and is recognized as a potent antitumor agent. Its mechanism involves affecting pyrimidine synthesis through the inhibition of thymidylate synthetase, leading to a depletion of intracellular dTTP pools.
  • Induces apoptosis and can serve as a chemical sensitizer.
  • Inhibits HIV.
  • Used as a reference standard in qualitative, quantitative, and methodological research experiments.
  • Applicable in techniques such as HPLC, GC, and MS.