Learn More
Medchemexpress LLC HY-107856 100mg Medchemexpress, 5-Fluorouridine CAS:316-46-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY107856100MG
Encompass_Preferred
Medchemexpress, HY-107856 100mg 5-Fluorouridine CAS:316-46-1 5-Fluorouridine is a metabolite of 5-fluorouracil with anticancer activity [1] [2] [3] [4] . 5-fluorouridine inhibits rRNA synthesis of human colon carcinoma cells [3] . 5-Fluorouridine exhibits cytotoxic effect on growth of L1210 cells with an IC 50 of 2 nM [4] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2303