Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluorouridine | 316-46-1 | MFCD00036832 | 100.0% | 262.19 g/mol | C9H11FN2O6 | 100 MG
SDP

Catalog No. 5000242429
Encompass_Preferred

5-Fluorouridine (CAS 316-46-1) is the analytical standard of 5-fluorouridine, a metabolite of 5-fluorouracil. It is intended for research and analytical applications and is supplied with defined purity and molecular properties to support accurate assays.

  • Analytical standard suitable for quantitative and qualitative assays.
  • High purity (99.97%) for reliable analytical results.
  • Molecular formula C9H11FN2O6; molecular weight 262.19 g/mol.
  • Available in 100 mg and solution/formulated pack sizes.
  • Metabolite that incorporates into RNA and induces apoptosis, useful for mechanistic studies.

Catalog No. 50-002-42429 Supplier Medchemexpress LLC Supplier No. HY107856R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluorouridine (CAS 316-46-1) is the analytical standard of 5-fluorouridine, a metabolite of 5-fluorouracil. It is intended for research and analytical applications and is supplied with defined purity and molecular properties to support accurate assays.

  • Analytical standard suitable for quantitative and qualitative assays.
  • High purity (99.97%) for reliable analytical results.
  • Molecular formula C9H11FN2O6; molecular weight 262.19 g/mol.
  • Available in 100 mg and solution/formulated pack sizes.
  • Metabolite that incorporates into RNA and induces apoptosis, useful for mechanistic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.