Learn More
Medchemexpress LLC 5-fluorouridine | 316-46-1 | MFCD00036832 | 262.19 | C9H11FN2O6 | 10 MG
SDP
Catalog No. 5000244504
Encompass_Preferred
5-Fluorouridine (standard) is an analytical reference standard intended for research and analytical applications. It is a metabolite of 5-fluorouracil and is reported to incorporate into RNA, exhibit antiproliferative activity, and induce apoptosis. This material is provided for laboratory research use only and is not validated for clinical or therapeutic use.
  • Analytical reference standard for method development and quantitation.
  • Supports identification and analytical comparison in research workflows.
  • Confirmed molecular formula C9H11FN2O6 and molecular weight 262.19.
  • CAS number 316-46-1 for unambiguous substance identification.
  • Certificate of analysis required for lot-specific purity and storage details.
Catalog No. 50-002-44504 Supplier Medchemexpress LLC Supplier No. HY107856R10MG
5-Fluorouridine (standard) is an analytical reference standard intended for research and analytical applications. It is a metabolite of 5-fluorouracil and is reported to incorporate into RNA, exhibit antiproliferative activity, and induce apoptosis. This material is provided for laboratory research use only and is not validated for clinical or therapeutic use.
  • Analytical reference standard for method development and quantitation.
  • Supports identification and analytical comparison in research workflows.
  • Confirmed molecular formula C9H11FN2O6 and molecular weight 262.19.
  • CAS number 316-46-1 for unambiguous substance identification.
  • Certificate of analysis required for lot-specific purity and storage details.