Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-fluorouridine | 316-46-1 | MFCD00036832 | 262.19 g·mol⁻¹ | C9H11FN2O6 | 50 MG
SDP

Catalog No. 5000244503
Encompass_Preferred

5-Fluorouridine analytical standard provided as a reference material for research and analytical applications. It is the ribonucleoside metabolite of 5-fluorouracil that incorporates into RNA and shows antiproliferative activity. Supplied as a solid for method development, calibration, and quality control; obtain the COA for purity and storage instructions.

  • Analytical standard for research and method development.
  • Suitable for calibration and quality control.
  • Ribonucleoside metabolite that incorporates into RNA.
  • Molecular formula C9H11FN2O6; molecular weight 262.19 g·mol⁻¹.
  • Pack size indicated as 50 mg; confirm with supplier documentation.

Catalog No. 50-002-44503 Supplier Medchemexpress LLC Supplier No. HY107856R50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-Fluorouridine analytical standard provided as a reference material for research and analytical applications. It is the ribonucleoside metabolite of 5-fluorouracil that incorporates into RNA and shows antiproliferative activity. Supplied as a solid for method development, calibration, and quality control; obtain the COA for purity and storage instructions.

  • Analytical standard for research and method development.
  • Suitable for calibration and quality control.
  • Ribonucleoside metabolite that incorporates into RNA.
  • Molecular formula C9H11FN2O6; molecular weight 262.19 g·mol⁻¹.
  • Pack size indicated as 50 mg; confirm with supplier documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.