Learn More
Bioworld 5-Fluorouridine, 1 g
Catalog No. 50256014
Encompass_Preferred
SpecificationsA proapoptotic and cytotoxic nucleoside analogSynonyms: 5-Fluorouracil 1β-D-ribofuranoside, FUrdShipped at ambient temperatureAppearance: SolidDensity : ~1.8 g/cm3 (Predicted)Melting Point: 182 °C
Catalog No. 50-256-014 Supplier Bioworld Supplier No. 406900173
SpecificationsA proapoptotic and cytotoxic nucleoside analogSynonyms: 5-Fluorouracil 1β-D-ribofuranoside, FUrdShipped at ambient temperatureAppearance: SolidDensity : ~1.8 g/cm3 (Predicted)Melting Point: 182 °C