Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical 5-(Fmoc-amino)-1-pentanol | 10 g | CAS 209115-33-3 | MDL MFCD00377789

Catalog No. 502362064
Encompass

5-(Fmoc-amino)-1-pentanol is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 209115-33-3
- MDL: MFCD00377789
- InChIKey: YNOWFUNORLDFTH-UHFFFAOYSA-N
- Molecular Weight: 325.408
- Molecular Formula: C20H23NO3
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (5-hydroxypentyl)carbamate
- SMILES: O=C(NCCCCCO)OCC1C2=CC=CC=C2C3=CC=CC=C31

Catalog No. 50-236-2064 Supplier STA Pharmaceutical Supplier No. STAAA1139010GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5-(Fmoc-amino)-1-pentanol is a Amino Acid reagent (Subcategory: Building Block) sold by WuXi TIDES. Offered in 10 g. Store at 4 °C. SDS available for reference.

Specifications
- CAS: 209115-33-3
- MDL: MFCD00377789
- InChIKey: YNOWFUNORLDFTH-UHFFFAOYSA-N
- Molecular Weight: 325.408
- Molecular Formula: C20H23NO3
- Purity: ≥95%
- Container Type: 60 mL HDPE
- Pack Size: 10 g
- Net Weight: 10 g
- Gross Weight: 25 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: (9H-fluoren-9-yl)methyl (5-hydroxypentyl)carbamate
- SMILES: O=C(NCCCCCO)OCC1C2=CC=CC=C2C3=CC=CC=C31

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.