Learn More
Gold Biotechnology Inc 5 FOA
SDP
Catalog No. 501532771
Encompass_Preferred
5-Fluoroorotic Acid (5-FOA) is widely used in the field of yeast molecular genetics The toxicity of 5-FOA to yeast cells with a functioning URA3 gene which encodes orotidine-5 -P decarboxylase (OMP decarboxylase) makes it especially useful for the selection and identification of mutant yeast strains 5-FOA is most often applied with Saccharomyces cerevisiae (URA3) although it has been used in other strains including Schizosaccharomyces pombe (URA4 and URA5) Candida albicans (URA3) and the bacterium E coli (pyrF) The use of 5-FOA in the yeast 2-hybrid system allows for the construction of activation domain hybrid libraries to identify protein-protein interactions
Catalog No. 50-153-2771 Supplier Gold Biotechnology Inc Supplier No. F2302.5
5-Fluoroorotic Acid (5-FOA) is widely used in the field of yeast molecular genetics The toxicity of 5-FOA to yeast cells with a functioning URA3 gene which encodes orotidine-5 -P decarboxylase (OMP decarboxylase) makes it especially useful for the selection and identification of mutant yeast strains 5-FOA is most often applied with Saccharomyces cerevisiae (URA3) although it has been used in other strains including Schizosaccharomyces pombe (URA4 and URA5) Candida albicans (URA3) and the bacterium E coli (pyrF) The use of 5-FOA in the yeast 2-hybrid system allows for the construction of activation domain hybrid libraries to identify protein-protein interactions