Learn More
Newcomer Supply Decalcifying Solution, Formic Acid 5%, Aqueous, 1 gallon
SDP
Catalog No. NC1578550
Encompass
Newcomer Supply Decalcifying Solution, Formic Acid, 5% Aqueous, provides a moderate rate of decalcification while maintaining excellent cellular morphology.  This solution is a general purpose decalcifier and is suitable for all bone specimen types from sternal or iliac crest bone marrow biopsies (light bone) to femoral head and long bone sections (compact bone). Shelf Life is 1 year from date of manufacture. Store at 15-30°C.
Catalog No. NC1578550 Supplier Newcomer Supply Supplier No. 1049C
Newcomer Supply Decalcifying Solution, Formic Acid, 5% Aqueous, provides a moderate rate of decalcification while maintaining excellent cellular morphology.  This solution is a general purpose decalcifier and is suitable for all bone specimen types from sternal or iliac crest bone marrow biopsies (light bone) to femoral head and long bone sections (compact bone). Shelf Life is 1 year from date of manufacture. Store at 15-30°C.