Learn More
Cayman Chemical 5-Formyl-2'-deoxyuridine, Purity: ≥95%, CAS: 4494-26-2, 10 mg
SDP
Catalog No. 502942321
Encompass_Preferred
A nucleoside derivative; inhibits the proliferation of BHK-21 cells and Ehrlich ascites tumor cells (IC50s = 4 and 6 µM, respectively); reduces virus yield in psuedorabies-infected BHK-21 cells at 1 mM; a product of oxidative DNA damage in vitro
Catalog No. 50-294-2321 Supplier Cayman Chemical Supplier No. 4030610 MG
A nucleoside derivative; inhibits the proliferation of BHK-21 cells and Ehrlich ascites tumor cells (IC50s = 4 and 6 µM, respectively); reduces virus yield in psuedorabies-infected BHK-21 cells at 1 mM; a product of oxidative DNA damage in vitro