Learn More
Medchemexpress LLC 5-formyl-2'-deoxyuridine | 4494-26-2 | MFCD01689820 | 99.6% | 256.21 g/mol | C10H12N2O6 | 5 MG
SDP
Catalog No. 5000282550
Encompass_Preferred
5-Formyl-2'-deoxyuridine is a nucleic acid derivative produced by oxidation of thymine and used in research to investigate oxidative DNA damage and repair. Supplied as a solid with high reported purity, it is suitable for analytical and biochemical applications, including chemical crosslinking studies.
  • Serves as a biomarker of oxidative DNA damage.
  • High purity (99.6%) for analytical use.
  • Solid form for convenient storage and handling.
  • Recommended storage: 4°C under inert gas; in solvent: -80°C (6 months), -20°C (1 month).
  • Identified by CAS number 4494-26-2 and formula C10H12N2O6.
Catalog No. 50-002-82550 Supplier Medchemexpress LLC Supplier No. HYW3414995MG
5-Formyl-2'-deoxyuridine is a nucleic acid derivative produced by oxidation of thymine and used in research to investigate oxidative DNA damage and repair. Supplied as a solid with high reported purity, it is suitable for analytical and biochemical applications, including chemical crosslinking studies.
  • Serves as a biomarker of oxidative DNA damage.
  • High purity (99.6%) for analytical use.
  • Solid form for convenient storage and handling.
  • Recommended storage: 4°C under inert gas; in solvent: -80°C (6 months), -20°C (1 month).
  • Identified by CAS number 4494-26-2 and formula C10H12N2O6.