Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formyl-2-furanboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F06115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0611-5G 5 g
F06111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0611-5G Supplier TCI America Supplier No. F06115G
Add to Cart
Add to Cart

Specifications

CAS 27329-70-0
Melting Point 151°C
Molecular Formula C5H5BO4
MDL Number MFCD01114696
Quantity 5 g
Synonym 2-formylfuran-5-boronic acid, 5-formylfuran-2-yl boronic acid, 5-formyl-2-furanboronic acid, 5-formylfuran-2-boronic acid, 5-formyl-2-furylboronic acid
InChI Key JUWYQISLQJRRNT-UHFFFAOYSA-N
SMILES B(C1=CC=C(O1)C=O)(O)O
IUPAC Name (5-formylfuran-2-yl)boronic acid
Molecular Weight (g/mol) 139.901
PubChem CID 2734355
Formula Weight 139.90
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.