Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formyl-2-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Catalog No. AAL19059MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19059MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL19059MD Supplier Thermo Scientific Chemicals Supplier No. L19059MD

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 127972-02-5
Melting Point ∼160°C
Molecular Formula C8H9BO4
Quantity 250 mg
MDL Number MFCD01319044
Beilstein 5334095
Sensitivity Air Sensitive
Synonym 2-methoxy-5-formylphenylboronic acid, 5-formyl-2-methoxyphenyl boronic acid, 5-formyl-2-methoxybenzeneboronic acid, 5-formyl-2-methoxy-phenyl boronic acid, 2-methoxy-5-formylbenzeneboronic acid, 3-borono-4-methoxybenzaldehyde, 3-dihydroxyboranyl-4-methoxybenzaldehyde, boronic acid, 5-formyl-2-methoxyphenyl, pubchem7991, acmc-209bd6
InChI Key NKKNXLPHCRLBDY-UHFFFAOYSA-N
SMILES COC1=CC=C(C=O)C=C1B(O)O
IUPAC Name (5-formyl-2-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 179.97
PubChem CID 2734367
Formula Weight 179.97
Percent Purity 98%
Chemical Name or Material 5-Formyl-2-methoxybenzeneboronic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.