Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formyl-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F08345G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0834-5G 5 g
F08341G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0834-5G Supplier TCI America Supplier No. F08345G
Add to Cart
Add to Cart

Specifications

CAS 127972-02-5
Color Yellow
Molecular Formula C8H9BO4
MDL Number MFCD01319044
Quantity 5 g
Synonym 2-methoxy-5-formylphenylboronic acid, 5-formyl-2-methoxyphenyl boronic acid, 5-formyl-2-methoxybenzeneboronic acid, 5-formyl-2-methoxy-phenyl boronic acid, 2-methoxy-5-formylbenzeneboronic acid
InChI Key NKKNXLPHCRLBDY-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)C=O)OC)(O)O
IUPAC Name (5-formyl-2-methoxyphenyl)boronic acid
Molecular Weight (g/mol) 179.966
PubChem CID 2734367
Formula Weight 179.97
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.