Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. F05495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F0549-5G 5 g
F05491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F0549-5G Supplier TCI America Supplier No. F05495G
Add to Cart
Add to Cart

Specifications

CAS 4347-33-5
Melting Point 135°C
Molecular Formula C5H5BO3S
Quantity 5 g
Synonym 5-formyl-2-thiopheneboronic acid, 5-formylthiophen-2-boronic acid, 5-formylthiophen-2-yl boronic acid, 5-formylthiophene-2-boronic acid, 2-formylthiophene-5-boronic acid
InChI Key DEQOVKFWRPOPQP-UHFFFAOYSA-N
SMILES B(C1=CC=C(S1)C=O)(O)O
IUPAC Name (5-formylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 155.962
PubChem CID 2773430
Formula Weight 155.96
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.