Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 5-Formyl-2-(trifluoromethyl)pyridine, 98%, [386704-12-7], F10292 175.108 C7H4F3NO

Catalog No. 501409196
Encompass_Preferred

Compound: 5-Formyl-2-(trifluoromethyl)pyridine

CAS#: 386704-12-7

MDL#: MFCD01862647

Background: A trifluoromethylated pyridine carbaldehyde used as a useful intermediate in organic synthesis, particularly in the preparation of pharmaceuticals and agrochemicals. The aldehyde group at the 3-position provides key reactivity for further derivatization.

Storage: Store at 2-8°C, under inert gas.

Catalog No. 50-140-9196 Supplier Frontier Specialty Chemicals Supplier No. F1029225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 5-Formyl-2-(trifluoromethyl)pyridine

CAS#: 386704-12-7

MDL#: MFCD01862647

Background: A trifluoromethylated pyridine carbaldehyde used as a useful intermediate in organic synthesis, particularly in the preparation of pharmaceuticals and agrochemicals. The aldehyde group at the 3-position provides key reactivity for further derivatization.

Storage: Store at 2-8°C, under inert gas.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.