Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-Formyl-4-methylthiophene-2-boronic acid 1,3-propanediol cyclic ester

Catalog No. SYX003411G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYX003411G 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYX003411G Supplier Sigma Aldrich Supplier No. SYX003411G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Undecanoyl chloride has been used in the synthesis of:
  • Chrysotrione b, 2-acylcyclopentene-1,3-dione derivative, isolated from the fruiting bodies of the basidiomycete hygrophorus chrysodon
  • 2-methylpentadecan-5-one
  • 4-ketotetradecanoic acid

Specifications

CAS 374537-98-1
Molecular Formula C9H11BO3S
Linear Formula C9H11BO3S
MDL Number MFCD02093958
Quantity 500 mg
Molecular Weight (g/mol) 210.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.