Learn More
Apexbio Technology LLC 5-Formyl-CTP 50ul (100 mM)
SDP
Catalog No. 5000088000
Encompass_Preferred
5-Formyl-CTP is a chemically modified nucleotide analog used to introduce formyl-cytidine residues into RNA transcripts It is designed to alter RNA secondary structures folding behaviors and interactions with RNA-binding proteins 5-Formyl-CTP exerts its biological impact by providing a formyl group at the 5-position of cytidine triphosphate facilitating the study of RNA structural dynamics and RNA-protein binding properties Additionally the formyl moiety enables chemical reactivity toward specific labeling reagents aiding in RNA detection labeling and structural analysis Based on these properties 5-Formyl-CTP holds research potential in biochemical and molecular biology applications including studies on RNA modifications structural biology and analytical labeling techniques
Catalog No. 50-000-88000 Supplier Apexbio Technology LLC Supplier No. B80530.05
5-Formyl-CTP is a chemically modified nucleotide analog used to introduce formyl-cytidine residues into RNA transcripts It is designed to alter RNA secondary structures folding behaviors and interactions with RNA-binding proteins 5-Formyl-CTP exerts its biological impact by providing a formyl group at the 5-position of cytidine triphosphate facilitating the study of RNA structural dynamics and RNA-protein binding properties Additionally the formyl moiety enables chemical reactivity toward specific labeling reagents aiding in RNA detection labeling and structural analysis Based on these properties 5-Formyl-CTP holds research potential in biochemical and molecular biology applications including studies on RNA modifications structural biology and analytical labeling techniques