Learn More
Apexbio Technology LLC 5-Formyl-dCTP(Synonyms: 5-Formyl-2'-deoxycytidine-5'-triphosphate, 5f-dCTP, fdCTP), 10ul (100 mM).
SDP
Catalog No. 5000060127
Encompass_Preferred
5-Formyl-dCTP is a modified cytosine-based deoxynucleoside triphosphate derivative capable of incorporation into DNA by DNA polymerase activity resulting in the formation of 5-formylcytosine (5fC) modified bases Presence of 5fC marks an intermediate step within active DNA demethylation pathways impacting epigenetic regulatory processes and gene expression It serves as a reagent in DNA modification and epigenomic studies facilitating investigations into the functional roles of 5fC in replication fidelity base excision repair mechanisms transcriptional regulation and chromatin remodeling dynamics
Catalog No. 50-000-60127 Supplier Apexbio Technology LLC Supplier No. B81170.01
Add to Cart
Add to Cart
5-Formyl-dCTP is a modified cytosine-based deoxynucleoside triphosphate derivative capable of incorporation into DNA by DNA polymerase activity resulting in the formation of 5-formylcytosine (5fC) modified bases Presence of 5fC marks an intermediate step within active DNA demethylation pathways impacting epigenetic regulatory processes and gene expression It serves as a reagent in DNA modification and epigenomic studies facilitating investigations into the functional roles of 5fC in replication fidelity base excision repair mechanisms transcriptional regulation and chromatin remodeling dynamics