Learn More
Apexbio Technology LLC 5-Formyl-dUTP(Synonyms: 5-Formyl-2'-deoxyuridine-5'-triphosphate, 5-Formyl-dUTP, f-dUTP, 5-fdUTP), 50ul (100 mM).
SDP
Catalog No. 5000060128
Encompass_Preferred
5-Formyl-dUTP is a nucleoside analog that disrupts DNA synthesis through incorporation into genomic DNA serving as a modifier of nucleic acid metabolism Upon integration this compound interferes with normal enzymatic activities involved in DNA replication and repair processes In biomedical research 5-Formyl-dUTP is utilized to investigate mechanisms underlying DNA damage responses and repair pathways By elucidating its impact on DNA physiology studies employing 5-Formyl-dUTP contribute to understanding the molecular events of genomic maintenance and inform therapeutic strategic developments for related human diseases
Catalog No. 50-000-60128 Supplier Apexbio Technology LLC Supplier No. B81200.05
5-Formyl-dUTP is a nucleoside analog that disrupts DNA synthesis through incorporation into genomic DNA serving as a modifier of nucleic acid metabolism Upon integration this compound interferes with normal enzymatic activities involved in DNA replication and repair processes In biomedical research 5-Formyl-dUTP is utilized to investigate mechanisms underlying DNA damage responses and repair pathways By elucidating its impact on DNA physiology studies employing 5-Formyl-dUTP contribute to understanding the molecular events of genomic maintenance and inform therapeutic strategic developments for related human diseases