Learn More
Selleck Chemical LLC 5-FU (5-Fluorouracil) S1209-10g
SDP
Catalog No. 503110969
Encompass_Preferred
5-FU (5-Fluorouracil) is a DNA/RNA synthesis inhibitor which interrupts nucleotide synthetic by inhibiting thymidylate synthase (TS) in tumor cells Fluorouracil induces apoptosis and can be used in the treatment of HIV
Catalog No. 50-311-0969 Supplier Selleck Chemical LLC Supplier No. S120910G
5-FU (5-Fluorouracil) is a DNA/RNA synthesis inhibitor which interrupts nucleotide synthetic by inhibiting thymidylate synthase (TS) in tumor cells Fluorouracil induces apoptosis and can be used in the treatment of HIV