Learn More
Medchemexpress LLC 5-(furan-2-yl)-N-(3-imidazol-1-ylpropyl)-1,2-oxazole-3-carboxamide | 909089-13-0 | MFCD14733081 | 99.7% | 286.29 g/mol | C14H14N4O3 | 200 MG
SDP
Catalog No. 5000213290
Encompass_Preferred
SKL2001 is a small-molecule agonist of the Wnt/β-catenin signaling pathway used in biomedical research. It stabilizes intracellular β-catenin by disrupting the Axin/β-catenin interaction and has reported anti-cancer activity. The compound is supplied as a high-purity solid and exhibits high solubility in DMSO, with available protocols for in vivo formulation.
Catalog No. 50-002-13290 Supplier Medchemexpress LLC Supplier No. HY101085200MG
SKL2001 is a small-molecule agonist of the Wnt/β-catenin signaling pathway used in biomedical research. It stabilizes intracellular β-catenin by disrupting the Axin/β-catenin interaction and has reported anti-cancer activity. The compound is supplied as a high-purity solid and exhibits high solubility in DMSO, with available protocols for in vivo formulation.