Learn More
US Plastic Corporation 5 Gallon/20 Liter Nalgene LDPE Round Carboy
SDP
Catalog No. NC1759638
Encompass
Dimensions: 282mm Diameter x 526mm High, Nominal Capacity to Brim: 5.5 Gallon/22.5 Liter, Cap Size: 83B.
Catalog No. NC1759638 Supplier US Plastic Corporation Supplier No. 72011
Dimensions: 282mm Diameter x 526mm High, Nominal Capacity to Brim: 5.5 Gallon/22.5 Liter, Cap Size: 83B.