Learn More
Km Custom Pack Jars
SDP
Catalog No. 501959323
Encompass_Preferred
The FisherbrandTM Glass Jars with Lids are made of clear soda lime glass and have a polypropylene screw cap lid with liner. They are available in a variety of sizes including 4oz, 8oz, 16oz, 32oz, 1 gallon, and 2 gallon. The jars can be used for a wide range of applications including storing dry goods, displaying items, and preparing food. The screw cap ensures an airtight seal. The polypropylene lid is durable, reusable, and withstands temperatures from -40°F to 200°F. Overall, these are high quality glass jars that provide versatile storage and display options.
Catalog No. 501959323 Supplier Km Custom Pack Supplier No. 5 GAL 1200G SANFRESH
The FisherbrandTM Glass Jars with Lids are made of clear soda lime glass and have a polypropylene screw cap lid with liner. They are available in a variety of sizes including 4oz, 8oz, 16oz, 32oz, 1 gallon, and 2 gallon. The jars can be used for a wide range of applications including storing dry goods, displaying items, and preparing food. The screw cap ensures an airtight seal. The polypropylene lid is durable, reusable, and withstands temperatures from -40°F to 200°F. Overall, these are high quality glass jars that provide versatile storage and display options.