Learn More
Uline Steel Pail with Lid - 5 Gallon, 5cs
Catalog No. NC1539849
Encompass
Extra strong for safe shipping of hazardous materials. Empty pails nest for storage. Recessed lid for easy stacking. Leakproof rubber gasket to ensure a tight seal.
Catalog No. NC1539849 Supplier Uline Supplier No. S7344CS
Extra strong for safe shipping of hazardous materials. Empty pails nest for storage. Recessed lid for easy stacking. Leakproof rubber gasket to ensure a tight seal.