Learn More
MSC Bel-Art Storage Tank, Polyethylene, Rectangle, 0.25 in Wall
Catalog No. 5000534425
Encompass
The product is a Bel-Art Polyethylene Storage Tank. It is designed for various storage needs, featuring a rectangular shape and a natural color. It is constructed with a durable 0.25 inch wall thickness and includes a 0.5 inch drain pipe.
  • Constructed from polyethylene for durability and chemical resistance.
  • Features a rectangular shape for efficient storage.
  • Equipped with a 0.25 inch wall thickness for robustness.
  • Includes a 0.5 inch drain pipe for convenient liquid transfer.
  • Natural color allows for visual inspection of contents.
  • Manufactured in the United States.
Catalog No. 50-005-34425 Supplier MSC Supplier No. 74338773
The product is a Bel-Art Polyethylene Storage Tank. It is designed for various storage needs, featuring a rectangular shape and a natural color. It is constructed with a durable 0.25 inch wall thickness and includes a 0.5 inch drain pipe.
  • Constructed from polyethylene for durability and chemical resistance.
  • Features a rectangular shape for efficient storage.
  • Equipped with a 0.25 inch wall thickness for robustness.
  • Includes a 0.5 inch drain pipe for convenient liquid transfer.
  • Natural color allows for visual inspection of contents.
  • Manufactured in the United States.