Learn More
US Plastic Corporation 5 Gallon Tamper Evident New Generation Container
SDP
Catalog No. NC1779656
Encompass
Width at widest point 12.62". Overall height with cover in place 16.04". Bottom outside diameter 10.27" Inside diameter at top 11.25". Inside height 15". Minimum cover wall thickness .090". Minimum pail wall thickness .090".
Catalog No. NC1779656 Supplier US Plastic Corporation Supplier No. 3741
Width at widest point 12.62". Overall height with cover in place 16.04". Bottom outside diameter 10.27" Inside diameter at top 11.25". Inside height 15". Minimum cover wall thickness .090". Minimum pail wall thickness .090".