Learn More
Justrite™ Dispensing Safety Can, 5-gal., Stainless Steel Hardware, Top Self-Close Brass Faucet, Flame Arrester
Catalog No. 19822441 Shop All Justrite Safety Group Products
Change view
Click to view available options
Color:
Red
Catalog No. Color
19-822-441 Red
1 options
Catalog No. 19-822-441 Supplier Justrite™ Supplier No. 14540
  • $576.00
    $374.65 / Each of 1
    Save $201.35
    35% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 5 Gallon Dispensing Safety Can with Stainless Steel Hardware, Brass Faucet and Flame Arrester is the choice of lab professionals for safely dispensing hazardous liquids into small flasks and beakers.
Specifications
Capacity (English) 5 gal.
Color Red
Model 14540
Includes Top faucet, self-close, brass
Material Polyethylene